Merck
  • Suppression of alcohol self-administration and reinstatement of alcohol seeking by melanin-concentrating hormone receptor 1 (MCH1-R) antagonism in Wistar rats.
Suppression of alcohol self-administration and reinstatement of alcohol seeking by melanin-concentrating hormone receptor 1 (MCH1-R) antagonism in Wistar rats.
Psychopharmacology (2010-07-16)
Andrea Cippitelli, Camilla Karlsson, Janice L Shaw, Annika Thorsell, Donald R Gehlert, Markus Heilig
RESUMEN
Melanin-concentrating hormone (MCH) is involved in regulation of appetitive behaviors as well as emotional reactivity and reward, behavioral domains relevant to alcohol addiction. We evaluated the effects of the non-peptide MCH1 receptor antagonist, GW803430 [6-(4-chloro-phenyl)-3-[3-methoxy-4-(2-pyrrolidin-1-yl-ethoxy)-phenyl]-3H-thieno[3,2-d]pyrimidin-4-one; 3-30 mg/kg, i.p.] on alcohol-related behaviors in Wistar rats. Ex vivo binding experiments demonstrated that the GW803430 dose range used resulted in high central MCH1 receptor occupancy. Alcohol self-administration was dose-dependently and potently suppressed, by approximately 80% at the highest dose. Reinstatement of alcohol-seeking induced by alcohol-associated cues was essentially eliminated. In contrast, reinstatement induced by footshock stress was not significantly altered. Taste preference for a quinine/saccharin solution, locomotor activity, and alcohol elimination were unaffected. Together, these observations support a specific involvement of the MCH system in mediating alcohol reward and cue-induced relapse to alcohol seeking. MCH1-R antagonism may constitute an attractive treatment target for alcohol use disorders.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
GW-803430, ≥98% (HPLC)