Merck
Regio- and stereo-selective oxidation of phenylbutane by rat liver.
Research communications in molecular pathology and pharmacology (1995-09-01)
M Takeshita, M Miura, Y Unuma, S Iwai, I Sato, T Arai, K Kosaka
RESUMEN
Regio- and stereo-selective oxidation of butylbenzene (1) has been examined in vitro by rat liver supernatant fraction(S-9). When phenylbutane (1 ) was incubated at 37 degrees C for 1 hr with S-9, asymmetric oxidation occurred regioselectively at benzylic and omega-1 positions to afford preferentially (R)- and (S)-alcohol (2, 4), respectively. This enzymatic propensity was the case for the production of 1, 3-diols. (1R, 3S)- or (1R, 3R)-Butanediols(3a, 3b) were also obtained at 87% and 27%, respectively. This oxidation was induced by phenobarbital (PB) or beta-napthoflavone(beta-NF), and significant sex-related differences in control and PB pre-treated rats have been observed. Since these oxidations were inhibited with SKF-525A and CO, it was inferred that cytochrome P-450 was responsible for the oxidation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Butylbenzene, ≥99%
Supelco
Butylbenzene, analytical standard