Merck
Gas chromatographic--mass spectrometric determination of amitriptyline and its major metabolites in human serum.
Journal of chromatography (1984-01-13)
R Ishida, T Ozaki, H Uchida, T Irikura
RESUMEN
A gas chromatographic--electron-impact ionization mass spectrometric method has been developed for the determination of amitriptyline (AMT) and its metabolites, nortriptyline (NT), 10-hydroxyamitriptyline (10-OH-AMT) and 10-hydroxynortriptyline (10-OH-NT) in human serum. The lower limit of detection was 2 ng/ml for all compounds except 5 ng/ml for 10-OH-NT. The calibration curves for AMT and 10-OH-AMT were linear up to 100 ng/ml, and up to 200 ng/ml for NT and 10-OH-NT. The accuracy of the assay in terms of coefficient of variation was less than 7%. The extraction efficiency was almost quantitative for all compounds except 60% for 10-OH-NT. Using this method, human serum samples which had been collected after oral administration of a single 50-mg dose of AMT were analyzed. Ratios of the conjugation of each metabolite were estimated, including AMT.