Merck
Pulsed excimer laser angioplasty of human cadaveric arteries.
Journal of vascular surgery (1986-02-01)
E M Farrell, L A Higginson, W S Nip, V M Walley, W J Keon
RESUMEN
Laser angioplasty has been limited by the lack of precise control of thermal and acoustic vascular injury. Pulsed excimer lasers, by contrast, have a capacity to affect target tissue without heat dispersion or damage to surrounding structures. The ablative properties of three excimer wavelengths, krypton fluoride (249 nm), xenon chloride (308 nm), and xenon fluoride (351 nm), were investigated with the use of fresh human cadaveric normal and atherosclerotic femoral arteries. Light and electron microscopy demonstrated clean cuts with histologically normal edges. There was no evidence of either thermal or acoustic damage with any of the wavelengths studied. The depth of ablation varied directly with the number of pulses and inversely with tissue density while the incision width remained constant. The excimer laser appears to offer significant advantages over its conventional counterparts for the ablation of atherosclerotic plaque.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Xenon difluoride, 99.99% trace metals basis