- Synthesis of 2'-O-methoxyethylguanosine using a novel silicon-based protecting group.
Synthesis of 2'-O-methoxyethylguanosine using a novel silicon-based protecting group.
The Journal of organic chemistry (2002-10-26)
Ke Wen, Suetying Chow, Yogesh S Sanghvi, Emmanuel A Theodorakis
PMID12398522
RESUMEN
A short and efficient synthesis of 2'-O-methoxyethylguanosine (8) is described. Central to this strategy is the development of a novel silicon-based protecting group (MDPSCl(2), 2) used to protect the 3',5'-hydroxyl groups of the ribose. Silylation of guanosine with 2 proceeded with excellent regioselectivity and in 79% yield. Alkylation of the 2'-hydroxyl group of 6 proceeded with methoxyethyl bromide and NaHMDS and afforded compound 7 in 85% yield, without any noticeable cleavage of the silyl protecting group and without the need to protect the guanine base moiety. Finally, deprotection of 7 was achieved using TBAF and produced 8 in 97% yield.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, purum, ≥98.0% (GC)