Merck
  • Development and validation of an HPLC method for the determination of six penicillin and three amphenicol antibiotics in gilthead seabream (Sparus Aurata) tissue according to the European Union Decision 2002/657/EC.
Development and validation of an HPLC method for the determination of six penicillin and three amphenicol antibiotics in gilthead seabream (Sparus Aurata) tissue according to the European Union Decision 2002/657/EC.
Food chemistry (2012-12-01)
Evaggelia N Evaggelopoulou, Victoria F Samanidou
RESUMEN
A confirmatory high performance liquid chromatography method for the determination of six penicillin antibiotics and three amphenicol antibiotics in gilthead seabream (Sparus Aurata) tissue was developed. Ampicillin (AMP), penicillin G (PG), penicillin V (PV), oxacillin (OXA), cloxacillin (CLO), dicloxacillin (DICLO), thiamphenicol (TAP), florfenicol (FFC) and chloramphenicol (CAP) were separated on an Inertsil, C(8) (250×4 mm, 5 μm) column by gradient elution with a mobile phase consisting of ammonium acetate 0.05 M and acetonitrile at 25°C. Diode array detection with monitoring at 225 nm (for the determination of AMP, PG, PV, TAP and FFC), 240 nm (for OXA, CLO and DICLO) and 278 nm (for CAP) was applied. Examined antibiotics were isolated from gilthead seabream tissue by liquid-liquid extraction and further clean-up was performed by solid phase extraction using Oasis HLB (200 mg/6 mL) cartridges. The developed method was fully validated in terms of selectivity, linearity, accuracy, precision, stability and sensitivity according to the European Union Decision 2002/657/EC.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Thiamphenicol
En este momento no podemos mostrarle ni los precios ni la disponibilidad