Merck
Influence of H2SO4 and ferric iron on Cd bioleaching from spent Ni-Cd batteries.
Waste management (New York, N.Y.) (2012-11-08)
Oksana Velgosová, Jana Kaduková, Renáta Marcinčáková, Pavol Palfy, Jarmila Trpčevská
RESUMEN
The paper is concerned with biohydrometallurgical methods of cadmium recovery from spent Ni-Cd batteries. Cd leaching efficiency from electrode material in different media (H(2)SO(4) and Fe(2)(SO(4))(3) solutions), at different Fe(III) concentrations and using the bacteria Acidithiobacillus ferrooxidans were investigated. The main aim of this study was to understand which from the bioleaching products (sulphuric acid or ferric sulphate) play a main role in the bioleaching process of Cd recovery. The influence of Fe ions on Cd leachability was confirmed. The best leaching efficiency of Cd (100%) was reached by bioleaching and also by leaching in Fe(2)(SO(4))(3) solution. The results of X-ray diffraction confirmed that no cadmium was present in solid residuum obtained after the Cd bioleaching as well as Cd leaching using solely ferric iron. The use of H(2)SO(4) solution resulted in the lowest efficiency of Cd leachability, the presence of hydroxides in electrode materials caused neutralization of the leaching solution and inhibition of Cd leaching.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Iron(III) sulfate hydrate, 97%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Iron(III) sulfate hydrate, BioReagent, suitable for plant cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad