Merck
Synthesis and asymmetric hydrogenation of (3E)-1-benzyl-3-[(2-oxopyridin-1(2H)-yl)methylidene]piperidine-2,6-dione.
Chemical communications (Cambridge, England) (2012-11-07)
Alexander A Bisset, Akira Shiibashi, Jasmine L Desmond, Allan Dishington, Teyrnon Jones, Guy J Clarkson, Takao Ikariya, Martin Wills
RESUMEN
The synthesis of (3E)-1-benzyl-3-[(2-oxopyridin-1(2H)-yl)methylidene]piperidine-2,6-dione 5 from N-benzylglutarimide was achieved in three steps. The asymmetric hydrogenation of 4 gave either the product of partial reduction (10) or full reduction (13), depending on the catalyst which was employed, in high ee in each case. Attempts at asymmetric transfer hydrogenation (ATH) of resulted in formation of a racemic product.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glutarimide, 98%