- Tuning efficiency of the 4-exo-trig cyclization by the electronic effect: ring closure of 3,3-difluoro-4-pentenyl carbon radicals and synthesis of a gem-difluorocyclobutane nucleoside.
Tuning efficiency of the 4-exo-trig cyclization by the electronic effect: ring closure of 3,3-difluoro-4-pentenyl carbon radicals and synthesis of a gem-difluorocyclobutane nucleoside.
Chemical communications (Cambridge, England) (2012-10-06)
Hiroki Kumamoto, Sachiko Kawahigashi, Hiromi Wakabayashi, Tomohiko Nakano, Tomoko Miyaike, Yasuyuki Kitagawa, Hiroshi Abe, Mika Ito, Kazuhiro Haraguchi, Jan Balzarini, Masanori Baba, Hiromichi Tanaka
PMID23038212
RESUMEN
4-exo-trig Cyclization reaction of a 4-pentenyl carbon radical containing the gem-difluoromethylene moiety adjacent to a radical accepting α,β-unsaturated ester was found to proceed efficiently to furnish a novel gem-difluorocyclobutane derivative. The cyclized product could be transformed into a gem-difluoromethylene analogue of oxetanocin T.