Merck
Modification of high saturated fat diet with n-3 polyunsaturated fat improves glucose intolerance and vascular dysfunction.
Diabetes, obesity & metabolism (2012-09-07)
K G Lamping, D W Nuno, L J Coppey, A J Holmes, S Hu, C L Oltman, A W Norris, M A Yorek
RESUMEN
The ability of dietary enrichment with monounsaturated fatty acid (MUFA), n-3 or n-6 polyunsaturated fatty acids (PUFAs) to reverse glucose intolerance and vascular dysfunction resulting from excessive dietary saturated fatty acids is not resolved. We hypothesized that partial replacement of dietary saturated fats with n-3 PUFA-enriched menhaden oil (MO) would provide greater improvement in glucose tolerance and vascular function compared to n-6 enriched safflower oil (SO) or MUFA-enriched olive oil (OO). We fed mice a high saturated fat diet (HF) (60% kcal from lard) for 12 weeks before substituting half the lard with MO, SO or OO for an additional 4 weeks. At the end of 4 weeks, we assessed glucose tolerance, insulin signalling and reactivity of isolated pressurized gracilis arteries. After 12 weeks of saturated fat diet, body weights were elevated and glucose tolerance was abnormal compared to mice on control diet (13% kcal lard). Diet substituted with MO restored basal glucose levels, glucose tolerance and indices of insulin signalling (phosphorylated Akt) to normal, whereas restoration was limited for SO and OO substitutions. Although dilation to acetylcholine was reduced in arteries from mice on HF, OO and SO diets compared to normal diet, dilation to acetylcholine was fully restored and constriction to phenylephrine was reduced in MO-fed mice compared to normal. We conclude that short-term enrichment of an ongoing high fat diet with n-3 PUFA rich MO, but not MUFA rich OO or n-6 PUFA rich SO, reverses glucose tolerance, insulin signalling and vascular dysfunction.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Safflower seed oil from Carthamus tinctorius seed