Merck
Highly enantioselective regiodivergent allylic alkylations of MBH carbonates with phthalides.
Journal of the American Chemical Society (2012-05-25)
Fangrui Zhong, Jie Luo, Guo-Ying Chen, Xiaowei Dou, Yixin Lu
RESUMEN
Phthalides were used for the first time in the allylic alkylation reactions with MBH carbonates for the creation of chiral 3,3-disubstituted phthalides. Highly enantioselective regiodivergent synthesis of γ-selective or β-selective allylic alkylation products was achieved by employing bifunctional chiral phosphines or multifunctional tertiary amine-thioureas as the catalyst, respectively. It was demonstrated that proper selection of catalysts and reaction conditions would differentiate an S(N)2'-S(N)2' pathway and an addition-elimination process, yielding different regioisomers of the allylic alkylation products in a highly enantiomerically pure form.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Phthalide, 98%