Merck
[Bradykinin-mediated angioedema secondary to angiotensin converting enzyme: initiate treatment from the prehospital phase].
Annales francaises d'anesthesie et de reanimation (2011-10-08)
N Javaud, O Fain, B Bernot, F Adnet, F Lapostolle
RESUMEN
We are reporting the case of a bradykinin-mediated angioedema, secondary to the angiotensin converting enzyme inhibitors, which delayed treatment could have unfavorably influence the vital prognostic of the patient. Initially, the patient had an isolated edema of the superior lip. Prehospital treatment included methylprednisolone, hydroxyzine and epinephrine. The patient was subsequently taken to the emergency department. His situation deteriorated. An edema of the cheeks and the tongue appeared. The transfer of the patient to an emergency department, specializing in kinin angioedema was organized, in order for the patient to receive specific treatments. After a subcutaneous injection of icatibant, the situation improved very rapidly, with a regression of the edema. This observation is consistent with the early use of the specific therapeutic in bradykinin-mediated angioedema. Any delay in administering the treatment can negatively impact the prognostic. The availability of such treatments should therefore be organized during the prehospital phase.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydroxyzine dihydrochloride, ≥97%