Merck
Pesticides removal from waste water by activated carbon prepared from waste rubber tire.
Water research (2011-06-15)
V K Gupta, Bina Gupta, Arshi Rastogi, Shilpi Agarwal, Arunima Nayak
RESUMEN
Waste rubber tire has been used for the removal of pesticides from waste water by adsorption phenomenon. By applying successive chemical and thermal treatment, a basically cabonaceous adsorbent is prepared which has not only a higher mesopore, macropore content but also has a favorable surface chemistry. Presence of oxygen functional groups as evidenced by FTIR spectra along with excellent porous and surface properties were the driving force for good adsorption efficiency observed for the studied pesticides: methoxychlor, methyl parathion and atrazine. Batch adsorption studies revealed maximum adsorption of 112.0 mg g(-1), 104.9 mg g(-1) and 88.9 mg g(-1) for methoxychlor, atrazine and methyl parathion respectively occurring at a contact time of 60 min at pH 2 from an initial pesticide concentration of 12 mg/L. These promising results were confirmed by column experiments; thereby establishing the practicality of the developed system. Effect of various operating parameters along with equilibrium, kinetic and thermodynamic studies reveal the efficacy of the adsorbent with a higher adsorption capacity than most other adsorbents. The adsorption equilibrium data obey Langmuir model and the kinetic data were well described by the pseudo-first-order model. Applicability of Bangham's equation indicates that diffusion of pesticide molecules into pores of the adsorbent mainly controls the adsorption process. Spontaneous, exothermic and random characteristics of the process are confirmed by thermodynamic studies. The developed sorbent is inexpensive in comparison to commercial carbon and has a far better efficiency for pesticide removal than most other adsorbents reported in literature.