Merck
Cu-catalyzed one-pot synthesis of unsymmetrical diaryl thioethers by coupling of aryl halides using a thiol precursor.
Organic letters (2011-01-29)
D J C Prasad, Govindasamy Sekar
RESUMEN
An efficient Cu-catalyzed one-pot approach for the synthesis of unsymmetrical diaryl thioethers using potassium ethyl xanthogenate as a thiol surrogate is developed. This new protocol avoids usage of intricate thiols and makes use of its easily available xanthate as a precursor, and thiol will be generated in situ to prepare the diaryl thioethers through a Cu-catalyzed double arylation. This strategy was further successfully utilized for the synthesis of symmetrical diaryl thioethers, aryl alkyl thioethers, and benzothiazoles.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Potassium ethyl xanthogenate, 96%
En este momento no podemos mostrarle ni los precios ni la disponibilidad