Merck
Occurrence and distribution of organophosphorus flame retardants and plasticizers in anthropogenically affected groundwater.
Journal of environmental monitoring : JEM (2010-12-07)
J Regnery, W Püttmann, C Merz, G Berthold
RESUMEN
Occurrence and distribution of chlorinated and non-chlorinated organophosphates in 72 groundwater samples from Germany under different recharge/infiltration conditions were investigated. Tris(2-chloro-1-methylethyl) phosphate (TCPP) and tris(2-chloroethyl) phosphate (TCEP) were the most frequently detected organophosphates in groundwater samples. Highest individual organophosphate concentrations (>0.1 µg L(-1)) were determined in groundwater polluted by infiltrating leachate and groundwater recharged via riverbank filtration of organophosphate-loaded recipients. In samples from springs and deep groundwater monitoring wells that are not affected by surface waters, organophosphate concentrations were mostly below the limit of detection. The occurrence (3-9 ng L(-1)) of TCPP and TCEP in samples from aquifers with groundwater ages between 20 and 45 years indicates the persistence of both compounds within the aquifer. At urban sites organophosphate-loaded precipitation, surface runoff, and leakage of wastewater influenced groundwater quality. For rural sites, where groundwater recharge is only influenced by precipitation, organophosphates were very rarely detectable in groundwater.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
TCPP, PESTANAL®, analytical standard