Merck
Trace detection of picloram using an electrochemical immunosensor based on three-dimensional gold nanoclusters.
Analytical biochemistry (2010-08-17)
Lijuan Chen, Guangming Zeng, Yi Zhang, Lin Tang, Danlian Huang, Can Liu, Ya Pang, Jie Luo
RESUMEN
Picloram, a herbicide widely used for broadleaf weed control, is persistent and mobile in soil and water with adverse health and environmental effects. It is important to develop a sensitive method for accurate detection of trace picloram in the environment. In this article, a type of ordered three-dimensional (3D) gold (Au) nanoclusters obtained by two-step electrodeposition using the spatial obstruction/direction of the polycarbonate membrane is reported. Bovine serum albumin (BSA)-picloram was immobilized on the 3D Au nanoclusters by self-assembly, and then competitive immunoreaction with picloram antibody and target picloram was executed. The horseradish peroxidase (HRP)-labeled secondary antibody was applied for enzyme-amplified amperometric measurement. The electrodeposited Au nanoclusters built direct electrical contact and immobilization interface with protein molecules without postmodification and positioning. Under the optimal conditions, the linear range for picloram determination was 0.001-10 μg/ml with a correlation coefficient of 0.996. The detection and quantification limits were 5.0 × 10(-4) and 0.0021 μg/ml, respectively. Picloram concentrations in peach and excess sludge supernatant extracts were tested by the proposed immunosensor, which exhibited good precision, sensitivity, selectivity, and storage stability.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4-Amino-3,5,6-trichloro-pyridine-2-carboxylic acid, BioReagent, suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Picloram, PESTANAL®, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad