Merck
Mesoporous beta-Ni(OH)2: synthesis and enhanced electrochemical performance.
Chemical communications (Cambridge, England) (2010-07-29)
Baojun Li, Man Ai, Zheng Xu
RESUMEN
Mesoporous beta-Ni(OH)(2) synthesized by a soft template method as a cathode material in a nickel based battery can provide higher specific discharge capacity in comparison with other micro/nanostructures.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nickel(II) hydroxide