Merck
  • Mono- and bidentate imidates of five-coordinate nickel(II) with macrocyclic ligands: spectroscopic and photophysical properties.
Mono- and bidentate imidates of five-coordinate nickel(II) with macrocyclic ligands: spectroscopic and photophysical properties.
Dalton transactions (Cambridge, England : 2003) (2010-05-25)
Rocío García-Bueno, M Dolores Santana, Gregorio Sánchez, Joaquín García, Gabriel García, José Pérez, Luis García
RESUMEN
The hydroxo-complexes [Ni(2)(mcN(3))(2)(mu-OH)](2)(PF(6))(2)] [mcN(3) = 2,4,4-trimethyl-1,5,9-triazacyclododec-1-ene (Me(3)-mcN(3)) or 2,4,4,9-tetramethyl-1,5,9-triazacyclododec-1-ene (Me(4)-mcN(3))] react with imides (Him) in 1 : 2 (succinimide, glutarimide, saccharine and 1,8-naphthalimide) or 1 : 1 (pyromellitic diimide) molar ratio, leading to the formation of the imidate complexes: [Ni(mcN(3))(H(2)O)(Im)](+) (Im = succi, gluti, sac), [Ni(mcN(3))(naphthi)](+) and [Ni(mcN(3))(H(2)O)](2)(mu-pyrdi)](2+). The single crystal structures of [Ni(Me(3)-mcN(3))(H(2)O)(sac)](PF(6)) (3a) and [Ni(Me(4)-mcN(3))(H(2)O)(sac)](sac) (3b) show that, in 3a, sac exhibits a N-monodentate coordination mode, while in 3b the coordination is through oxygen. X-Ray structure of [Ni(Me(4)-mcN(3))(naphthi)](PF(6)) (4b) shows N,O-bonded 1,8-naphthalimidate ligand. The VT (1)H NMR study, carried out on this complex, suggests that the rotation around the Ni-N bond is hindered by ca. 11.6 kcal mol(-1). These species exhibit blue luminescence which arises from a mixed MLCT and LC excited state, and these properties may be useful for structural predictions in the solid state of complexes for which crystal structures have not been established.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Pyromellitic diimide, 97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad