Merck
  • Identification of covalent binding sites of ethyl 2-cyanoacrylate, methyl methacrylate and 2-hydroxyethyl methacrylate in human hemoglobin using LC/MS/MS techniques.
Identification of covalent binding sites of ethyl 2-cyanoacrylate, methyl methacrylate and 2-hydroxyethyl methacrylate in human hemoglobin using LC/MS/MS techniques.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-05-18)
Marina C Jeppsson, Harriet Mörtstedt, Giovanni Ferrari, Bo A G Jönsson, Christian H Lindh
RESUMEN
Acrylates are used in vast quantities, for instance in paints, adhesive glues, molding. They are potent contact allergens and known to cause respiratory hypersensitivity and asthma. Here we study ethyl 2-cyanoacrylate (ECA), methyl methacrylate (MMA) and 2-hydroxyethyl methacrylate (HEMA). There are only limited possibilities to measure the exposure to acrylates, especially for biological monitoring. The aim of the present study was to investigate the chemical structures of adducts formed after reaction of hemoglobin (Hb) with ECA, MMA, and HEMA. This information may be used to identify adducted Hb peptides for biological monitoring of exposure to acrylates. Hb-conjugates with ECA, MMA, and HEMA were synthesized in vitro. The conjugates were digested by trypsin and pronase E. Adducted peptides were characterized and analyzed by liquid chromatography and nano electro spray/hybrid quadrupole time-of-flight mass spectrometry (MS) as well as tandem quadrupole MS. The search for the adducted peptides was facilitated by visualizing the MS data by different computer programs. The results showed that ECA binds covalently to cysteines at the 104 position in the α and the position 112 in the β-chains in Hb. MMA and HEMA bound to all the cysteines in both chains, Cys(104) in the α-chain and Cys(93) and 112 in the β-chain. The full-length spectra of in un-digested Hb confirmed this binding pattern. There was no reaction with N-acetyl-L-lysine at physiological pH. The adducted peptides were possible to measure using LC/MS/MS in selected reaction monitoring mode. These peptides may be used for biological monitoring of exposure to ECA, MMA and HEMA.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethyl 2-cyanoacrylate, liquid