Merck
Photochemical decomposition of perfluorooctanoic acid in aqueous periodate with VUV and UV light irradiation.
Journal of hazardous materials (2010-04-21)
M H Cao, B B Wang, H S Yu, L L Wang, S H Yuan, J Chen
RESUMEN
The photochemical decomposition of perfluorooctanoic acid (PFOA) in aqueous periodate (IO(4)(-)) was investigated under two types of low-pressure mercury lamps: one emits at 254nm light (UV light) and the other emits both 254 nm and 185 nm light (VUV light). PFOA decomposed efficiently under VUV light irradiation while it decomposed poorly under UV light irradiation. The addition of IO(4)(-) significantly increased the rate of decomposition and defluorination of PFOA irradiated with UV light whereas it decreased both processes under VUV irradiation. Reactive radical (IO(3)) generated by photolysis of IO(4)(-) initiated the oxidation of PFOA in UV process. Aquated electrons (e(aq)(-)), generated from water homolysis, scavenged IO(4)(-) resulting in decrease of reactive radical species production and PFOA decomposition. The shorter-chain perfluorocarboxylic acids (PFCAs) formed in a stepwise manner from long-chain PFCAs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium iodate, ≥99%
Sigma-Aldrich
Sodium iodate, puriss. p.a., ≥99.5% (RT)