Merck
Transglycosylation activity of cellobiohydrolase I from Trichoderma longibrachiatum on synthetic and natural substrates.
Biochimica et biophysica acta (1991-04-09)
A V Gusakov, O V Protas, V M Chernoglazov, A P Sinitsyn, G V Kovalysheva, O V Shpanchenko, O V Ermolova
RESUMEN
Using 4-methylumbelliferyl (MUF) beta-D-cellobioside as a substrate, the ability of cellobiohydrolase I from Trichoderma longibrachiatum to catalyze transglycosylation has been demonstrated. At substrate concentrations greater than 2 mM, the formation of MUF-tetrasaccharide was detected using HPLC. In the course of enzymatic reaction, a concentration of the transglycosylation product passed through a maximum, since at later stages of the reaction the product was further hydrolyzed. At MUF-beta-D-cellobioside concentrations of 2-10 mM, the maximum weight content of MUF-tetrasaccharide amounted to 1-4% of the total content of saccharides. In the reaction system, containing 2.5 mM MUF-beta-D-cellobioside and 10 mM MUF-beta-D-glucoside, MUF-trisaccharide was formed as the main transglycosylation product. In hydrolysis of natural substrates (cellulose and cellotriose) in the presence of MUF-beta-D-glucoside a formation of MUF-trisaccharide was also observed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4-Methylumbelliferyl β-D-cellobioside, glucanase substrate
En este momento no podemos mostrarle ni los precios ni la disponibilidad