Merck
Chemical constituents from the fruits of Hippophae rhamnoides.
Natural product research (2009-10-08)
Rui-Xia Zheng, Xu-Dong Xu, Ze Tian, Jun-Shan Yang
RESUMEN
Ten compounds were isolated from the fruits of Hippophae rhamnoides. On the basis of spectroscopic and chemical methods, the structures of these compounds were elucidated as hippophae cerebroside (1), oleanolic acid (2), ursolic acid (3), 19-alpha-hydroxyursolic acid (4), dulcioic acid (5), 5-hydroxymethyl-2-furancarbox-aldehyde (6), cirsiumaldehyde (7), octacosanoic acid (8), palmitic acid (9) and 1-O-hexadecanolenin (10). Among them, 1 was a new compound, and 4-7 and 10 were obtained from the genus for the first time.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Octacosanoic acid, synthetic, ≥98%