Merck
  • 1,3-Dioxolane-based ligands as rigid analogues of naftopidil: structure-affinity/activity relationships at alpha1 and 5-HT1A receptors.
1,3-Dioxolane-based ligands as rigid analogues of naftopidil: structure-affinity/activity relationships at alpha1 and 5-HT1A receptors.
ChemMedChem (2009-01-20)
Claudia Sorbi, Silvia Franchini, Annalisa Tait, Adolfo Prandi, Rossella Gallesi, Piero Angeli, Gabriella Marucci, Lorenza Pirona, Elena Poggesi, Livio Brasili
RESUMEN
Conformational restriction of naftopidil proved to be compatible with binding at alpha(1) adrenoceptor subtypes and 5-HT receptor 1A (5-HT(1A)), and led to the discovery of a new class of ligands with a 1,3-dioxolane (1,3-oxathiolane, 1,3-dithiolane) structure. Compound 7 shows the highest affinity toward alpha(1a) and alpha(1d) adrenoceptor subtypes (pK(i) alpha(1a) = 9.58, pK(i) alpha(1d) = 9.09) and selectivity over 5-HT(1A) receptors (alpha(1a)/5-HT(1A) = 100, alpha(1d)/5-HT(1A) = 26). In functional experiments it behaves as a potent competitive alpha(1a) and alpha(1d) adrenoceptor antagonist (pK(b) alpha(1A) = 8.24, pK(b) alpha(1D) = 8.14), whereas at 5-HT(1A) receptors it is a potent partial agonist (pD(2) = 8.30). Compounds 8 and 10 display high affinity (pK(i) = 8.29 and 8.26, respectively) and selectivity for 5-HT(1A) (5-HT(1A)/alpha(1) = 18 and 10). In functional experiments at the 5-HT(1A) receptor, compound 8 appears to be neutral antagonist (pK(b) = 7.29), whereas compound 10 is a partial agonist (pD(2) = 6.27). Therefore, 1,3-dioxolane-based ligands are a versatile class of compounds useful for the development of more selective ligands for one (alpha(1)) or the other (5-HT(1A)) receptor system.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,3-Dioxolano, ReagentPlus®, contains ~75 ppm BHT as inhibitor, 99%
Sigma-Aldrich
1,3-Dioxolano, anhydrous, contains ~75 ppm BHT as inhibitor, 99.8%