Merck
Synthesis and biological evaluation of a series of tangeretin-derived chalcones.
Bioorganic & medicinal chemistry letters (2008-11-18)
Jérôme Quintin, Julie Desrivot, Sylviane Thoret, Patrick Le Menez, Thierry Cresteil, Guy Lewin
RESUMEN
A series of chalcones polyoxygenated on the ring A (with pentamethoxy or 2'-hydroxy-3',4',5',6'-tetramethoxy substitution patterns) was synthesized from tangeretin, a natural Citrus flavonoid. These chalcones were evaluated for their antiproliferative, activation of apoptosis, inhibition of tubulin assembly and antileishmanial activities. Comparison with the reference analogous 3',4',5'-trimethoxylated chalcones showed that such peroxygenated substitution patterns on the ring A were less beneficial to these activities.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tangeretin, ≥95% (HPLC)