Merck
Immune-mediated inflammatory diseases (IMIDs) and biologic therapy: a medical revolution.
Postgraduate medical journal (2007-04-04)
Annabel Kuek, Brian L Hazleman, Andrew J K Ostör
RESUMEN
Targeted biologic therapies have revolutionised treatment of immune-mediated inflammatory diseases (IMIDs) due to their efficacy, speed of onset and tolerability. The discovery that clinically unrelated conditions, such as rheumatoid arthritis and Crohn's disease, share similar immune dysregulation has led to a shift in the management of IMIDs from one of organ-based symptom relief to mechanism-based treatment. The fact that anticytokine therapy has been effective in treating multiple orphan inflammatory conditions confirms the IMID paradigm. In this review we examine the biologic agents currently licensed for use in the US and Europe: infliximab, etanercept, adalimumab, rituximab, abatacept, anakinra, alefacept and efalizumab. We also discuss the rationale behind the management of IMIDs using rheumatoid arthritis, Crohn's disease, psoriasis and psoriatic arthritis as examples. For the medical profession, IMID represents a breakthrough in the way pathology is classified. In this burgeoning era of biologic therapy the prospect of complete disease remission is conceivable.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Ethoxyaniline, 98%