Merck
Construction of Lactobacillus plantarum strain with enhanced L-lysine yield.
Journal of applied microbiology (2007-02-21)
M N Cahyanto, H Kawasaki, M Nagashio, K Fujiyama, T Seki
RESUMEN
To enhance L-lysine secretion in Lactobacillus plantarum. An S-2-aminoethyl-L-cystein (AEC)-resistant mutant of L. plantarum was isolated, and it produced L-lysine at considerably higher level than the parent strain. Aspartokinase in the mutant has been desensitized to feedback inhibition by L-lysine. The nucleotide sequence analysis of thrA2 that codes for aspartokinase in the mutant predicted a substitution of glutamine to histidine at position 421. L-Lysine-insensitive aspartokinase, together with aspartate semialdehyde dehydrogenase, dihydrodipicolinate synthase, and dihydrodipicolinate reductase genes, was cloned from L. plantarum DNA to a shuttle vector, pRN14, and the genes were then transformed individually into the AEC-resistant mutant and the parent strain. The overexpression of the genes led to the increase in the activity of enzymes they encode in vitro. However, only the strain overexpressing aspartokinase or dihydrodipicolinate synthase produced more L-lysine. The desensitization of aspartokinase to L-lysine in L. plantarum led to the overproduction of L-lysine. The overexpression of L-lysine-insensitive aspartokinase or dihydrodipicolinate synthase enhanced L-lysine secretion in L. plantarum. The use of the L-lysine-overproducing strain of L. plantarum in food or feed fermentation may increase the L-lysine content of fermented products.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
S-(2-Aminoethyl)-L-cysteine hydrochloride, ≥98% (TLC)