Merck
  • Novel affinity chromatographic system for the single-step purification of glycosaminoglycans from complex systems using volatile buffers.
Novel affinity chromatographic system for the single-step purification of glycosaminoglycans from complex systems using volatile buffers.
Journal of chromatography (1991-04-19)
B A Hodson, D S Pepper, J Dawes
RESUMEN
A new system for the isolation and purification of glycosaminoglycans (GAGs) and related molecules from complex systems such as plasma is described. Affinity chromatography which exploits the very high affinity between the polymeric base Polybrene and sulphated polysaccharides was used. A novel volatile buffer system composed of ammonium formate and formic acid, which allows the complete recovery of samples, was developed, and elution conditions were optimised for the separation and purification of GAGs of different charge densities. Using this system the losses associated with dialysis and desalting, frequently necessary preliminaries to further analysis, are avoided.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Formato de amonio, BioUltra, ≥99.0% (calc. based on dry substance, NT)