Merck
  • Glucagon-like peptide-1, corticotropin-releasing hormone, and hypothalamic neuronal histamine interact in the leptin-signaling pathway to regulate feeding behavior.
Glucagon-like peptide-1, corticotropin-releasing hormone, and hypothalamic neuronal histamine interact in the leptin-signaling pathway to regulate feeding behavior.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2005-05-17)
Koro Gotoh, Koji Fukagawa, Tomiyo Fukagawa, Hitoshi Noguchi, Tetsuya Kakuma, Toshiie Sakata, Hironobu Yoshimatsu
RESUMEN
Glucagon-like peptide-1 (GLP-1), corticotropin-releasing hormone (CRH), and hypothalamic neuronal histamine suppress food intake, a target of leptin action in the brain. This study examined the interactions of GLP-1, CRH, and histamine downstream from the leptin-signaling pathway in regulating feeding behavior. Infusion of GLP-1 into the third cerebral ventricle (i3vt) at a dose of 1 mug significantly decreased the initial 1 h cumulative food intake in rats as compared with phosphate-buffered saline (PBS) controls. The GLP-1-induced suppression of feeding was partially attenuated by intraperitoneal pretreatment with alpha-fluoromethylhistidine (FMH), a specific suicide inhibitor of histidine decarboxylase, which depletes hypothalamic neuronal histamine. Pretreatment with alpha-helical CRH (10 microg/rat, i3vt), a nonselective CRH antagonist, abolished the GLP-1-induced suppression of feeding completely. I3vt infusion of GLP-1 increased the CRH content and histamine turnover assessed using the pargyline-induced accumulation of tele-methyl histamine (t-MH), a major metabolite of neuronal histamine, in the hypothalamus. The central infusion of CRH also induced the increase of histamine turnover and CRH receptor type 1 was localized on the cell body of histamine neuron. Pretreatment with exendin(9-39), a GLP-1 receptor antagonist, attenuated the leptin-induced increase in CRH content of the hypothalamus. Finally, i3vt infusion of leptin also increased histamine turnover in the hypothalamus. Pretreatment with exendin(9-39), alpha-helical CRH or both antagonists attenuated the leptin-induced responses of t-MH levels in the hypothalamus. These results suggest that CRH or hypothalamic neuronal histamine mediates the GLP-1-induced suppression of feeding behavior, that CRH mediates GLP-1 signaling to neuronal histamine and that a functional link from GLP-1 to neuronal histamine via CRH constitutes the leptin-signaling pathway regulating feeding behavior.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1-Methylhistamine dihydrochloride, ≥98% (TLC), powder