Merck
Mouse spermine oxidase: a model of the catalytic cycle and its inhibition by N,N1-bis(2,3-butadienyl)-1,4-butanediamine.
Biochemical and biophysical research communications (2004-08-18)
Andrea Bellelli, Stefano Cavallo, Laura Nicolini, Manuela Cervelli, Marzia Bianchi, Paolo Mariottini, Massimo Zelli, Rodolfo Federico
RESUMEN
Spermine oxidase (SMO) is a recently described flavoenzyme belonging to the class of polyamine oxidases (PAOs) and participating in the polyamine metabolism in animal cells. In this paper we describe the expression, purification, and characterization of the catalytic properties of a recombinant mouse SMO (mSMO). The purified enzyme has absorbance peaks at 457nm (epsilon=11mM(-1)cm(-1)) and 378nm, shows a molecular mass of approximately 63kDa, and has K(m) and k(cat) values of 170microM and 4.8s(-1), using spermine as substrate; it is unable to oxidize other free or acetylated polyamines. The mechanism-based PAO inhibitor N,N(1)-bis(2,3-butadienyl)-1,4-butanediamine (MDL72,527) acts as a competitive inhibitor of mSMO, with an apparent dissociation constant K(i)=63microM. If incubated for longer times, MDL72,527 yields irreversible inhibition of the enzyme with a half-life of 15min at 100microM MDL72,527. The mMSO catalytic mechanism, investigated by stopped flow, is consistent with a simple four-step kinetic scheme.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
MDL 72527, ≥98% (HPLC)