Merck
Hypothalamus-brain stem circuitry responsible for vagal efferent signaling to the pancreas evoked by hypoglycemia in rat.
Journal of neurophysiology (2003-12-03)
Xiaoyin Wu, Jun Gao, Jin Yan, Chung Owyang, Ying Li
RESUMEN
Circulating glucose levels significantly affect vagal neural activity, which is important in the regulation of pancreatic functions. Little is known about the mechanisms involved. This study investigates the neural pathways responsible for hypoglycemia-induced vagal efferent signaling to the pancreas and identifies the neurotransmitters involved. Vagal pancreatic efferent nerve activities were recorded in anesthetized rats. Insulin-induced hypoglycemia, a decrease of blood glucose levels from 114 +/- 5 to 74 +/- 6 mg dl(-1), stimulated an increase in pancreatic efferent nerve firing from a basal rate of 1.1 +/- 0.3 to 19 +/- 3 impulses 30 s(-1). In contrast, vagal primary afferent neuronal discharges recorded in the nodose ganglia were unaltered by systemic hypoglycemia. Vagal afferent rootlet section plus splanchnicotomy had no effect on hypoglycemia-induced vagal efferent firing, suggesting a central site of action. Decerebration reduced the increase in nerve firing stimulated by hypoglycemia from 21 +/- 4 to 9.6 +/- 2 impulses 30 s(-1). Chemical ablation of the lateral hypothalamic area, but not the arcuate nucleus, inhibited pancreatic nerve firing evoked by hypoglycemia. Microinjection of the orexin-A receptor antagonist SB-334867 into the dorsal motor nucleus of the vagus (DMV) inhibited pancreatic nerve firing evoked by insulin-induced hypoglycemia by 56%. In contrast, injection of orexin-A (20 pmol) into the DMV elicited a 30-fold increase in pancreatic nerve firing. We concluded that systemic hypoglycemia stimulates pancreatic efferent nerve firing through a central mechanism. Full expression of pancreatic nerve activities during hypoglycemia requires both the forebrain and the brain stem. In addition to activating neurons in the brain stem, central neuroglucopenia activates subpopulations of neurons in the lateral hypothalamic area that contain orexin. The released orexin acts on DMV neurons to stimulate pancreatic efferent nerve activities and thus regulate pancreatic functions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
5-Thio-D-glucose, ≥98.0% (HPLC)