Merck
  • Intermolecular transition metal-catalyzed [4 + 2 + 2] cycloaddition reactions: a new approach to the construction of eight-membered rings.
Intermolecular transition metal-catalyzed [4 + 2 + 2] cycloaddition reactions: a new approach to the construction of eight-membered rings.
Journal of the American Chemical Society (2002-07-26)
P Andrew Evans, John E Robinson, Erich W Baum, Aleem N Fazal
RESUMEN
Transition metal-catalyzed cycloaddition reactions represent powerful methods for the construction of complex polycyclic systems. We have developed a new intermolecular metal-catalyzed [4 + 2 + 2] cycloaddition of heteroatom-tethered enyne derivatives with 1,3-butadiene. This study demonstrates that excellent selectivity can be obtained for the heterocycloaddition adducts through the judicious choice of silver salt. The development of the tandem rhodium-catalyzed allylic substitution [4 + 2 + 2] cycloaddition provides a convenient three-component coupling that circumvents the prior formation of the enyne derivative. Finally, the introduction of a stereogenic center at C-2 leads to a diastereoselective cycloaddition, which provides a powerful new method for the construction of bicyclic octanoid ring systems applicable to target directed synthesis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tris(triphenylphosphine)rhodium(I) chloride, 99.9% trace metals basis
Sigma-Aldrich
Tris(triphenylphosphine)rhodium(I) chloride