Merck
Angiotensin converting enzyme inhibition prevents trophic and hypertensive effects of an antagonist of adenosine receptors.
European journal of pharmacology (2002-05-15)
Teresa Sousa, Manuela Morato, António Albino-Teixeira
RESUMEN
The continuous infusion of 1,3-dipropyl-8-sulfophenylxanthine (DPSPX), a non-selective antagonist of adenosine receptors, causes hypertension and marked cardiovascular structural changes in Wistar rats. Adenosine inhibits noradrenaline and renin release. We investigated the effects of sympathetic denervation, evaluated renin activity and the influence of angiotensin converting enzyme inhibition in DPSPX-treated rats. Captopril was given (30 or 100 mg kg(-l) day(-l); p.o.) from day -l to day 28. On day 0, constant infusions of DPSPX (90 microg kg(-l) h(-l); i.p.) or vehicle were started. On day 28, fragments of the left ventricle, mesenteric and tail arteries were processed for morphological studies. Plasma renin activity was increased in DPSPX-treated animals. Sympathetic denervation delayed and partially prevented blood pressure rise. Angiotensin converting enzyme inhibition prevented DPSPX-induced hypertension and morphological changes. Our results, although pointing to the involvement of the sympathetic system, suggest that other mechanisms are involved. We could not differentiate between the trophic and anti-hypertensive effects of angiotensin converting enzyme inhibition.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,3-Dipropyl-8-(p-sulfophenyl)xanthine, powder