Merck
Molecular weight of dissolved organic matter-napropamide complex transported through soil columns.
Journal of environmental quality (2002-04-05)
C F Williams, J Letey, W J Farmer
RESUMEN
Soil-derived dissolved organic matter (DOM) has been shown to form stable complexes with the herbicide napropamide [2-(alpha-naphthoxy-N,N-diethylpropionamide] capable of enhancing the transport of napropamide through soil columns. Two soils, one containing sewage sludge-derived organic matter (SS) and the other having only natural organic matter (NoSS) were treated with napropamide and allowed to dry to promote complex formation. Soil columns were prepared by packing a 10-cm layer of untreated, dry, sieved soil followed by an overlying 5-cm layer of napropamide-treated soil. Columns were irrigated and the effluent collected and placed in dialysis chambers. After equilibration napropamide concentrations were determined on both sides of the membrane and complex and quantified based on the amount of napropamide unable to cross the membrane. it was found that for the SS soil 7% and for the NoSS 2.4% of the applied napropamide underwent facilitated transport. In addition, most of the complex transported through the columns had a molecular weight between 500 and 1000 Daltons (Da). The solutions from the SS soil were also found to have formed at least two distinct complexes that were resolved after passing through the untreated soil layer. The results obtained were in agreement with other published results and the techniques used offer a way to separate and concentrate DOM complexes from column effluents for further characterization.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Napropamid, PESTANAL®, analytical standard