Merck
  • Human V gamma 2V delta 2 T cells produce IFN-gamma and TNF-alpha with an on/off/on cycling pattern in response to live bacterial products.
Human V gamma 2V delta 2 T cells produce IFN-gamma and TNF-alpha with an on/off/on cycling pattern in response to live bacterial products.
Journal of immunology (Baltimore, Md. : 1950) (2001-11-21)
L Wang, H Das, A Kamath, J F Bukowski
RESUMEN
Whereas cytokine production in alphabeta T cells is rapidly regulated by exposure to peptide Ag, the mechanisms regulating cytokine production by gammadelta T cells are unknown. In this study, we demonstrate that human Vgamma2Vdelta2 T cells produce IFN-gamma and TNF-alpha as early as 2 h after Ag exposure, and that they produce these cytokines in a dose- and time- dependent manner in response to stimulation with a live bacterial product, iso-butylamine (IBA), but not to dead bacteria or LPS. gammadelta T cells began, ceased, and then resumed IFN-gamma and TNF-alpha generation in an on/off/on cycling pattern, both in vitro and in vivo, depending on the presence or absence of IBA. IFN-gamma and TNF-alpha, whose optimum production was dependent on IBA-stimulated gammadelta T cells, were critical for monocyte-mediated killing of Escherichia coli. By limiting cytokine production to periods of direct contact with live bacteria, gammadelta T cells focus their resources at the site of infection, while limiting systemic immunopathology. Thus, human gammadelta T cells may mediate innate resistance to extracellular bacteria via tightly regulated cytokine production without necessarily expanding in number.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Isobutylamine, 99%