Merck
Is salivary histatin 5 a metallopeptide?
Biochimica et biophysica acta (2001-05-09)
H Gusman, U Lendenmann, J Grogan, R F Troxler, F G Oppenheim
RESUMEN
Histatins are small histidine-rich salivary polypeptides which exhibit antimicrobial activity against Candida albicans. This antimicrobial activity has been ascribed in part to a high content of basic amino acids. However, unlike most other antimicrobial proteins histatins have a high content of histidine, tyrosine and acidic amino acids known to participate in metal ion coordination. This study was conducted to test whether histatin 5 could bind zinc and copper which are metals present in salivary secretions and whole saliva. Physical binding parameters and spectral properties of zinc- and copper-histatin complexes were investigated in order to obtain direct evidence of these interactions. A spectrophotometric competition assay using the metallochromic indicator murexide showed that histatin 5 dissociates metal indicator complexes containing zinc or copper ions. Absorption spectra of histatin 5 at increasing copper chloride concentrations resulted in higher absorbance in the 230-280 nm wavelength range and this spectral change was saturated at a peptide:metal molar ratio of approx. 1:1. A corresponding band was observed in the visible range of the spectrum with a maximum and molar extinction coefficient corresponding to that of copper binding to an ATCUN motif. Quantitative assessment of zinc and copper binding to histatin 5 using isothermal titration calorimetry revealed at least one high affinity site for each metal, with binding constants of 1.2x10(5) and 2.6x10(7) M(-1), respectively. These results indicate that histatin 5 exhibits metallopeptide-like properties. The precise biological significance of this has not yet been established but histatins may contribute significantly to salivary metal binding capacity.