Merck
A one-flask synthesis of Weinreb amides from chiral and achiral carboxylic acids using the deoxo-fluor fluorinating reagent.
Organic letters (2000-12-12)
A R Tunoori, J M White, G I Georg
RESUMEN
[structure] The reagent [bis(2-methoxyethyl)amino]sulfur trifluoride (Deoxo-Fluor reagent) converts carboxylic acids to the corresponding acid fluorides, which then react with N,N-dimethylhydroxylamine to give the corresponding Weinreb amides in high yields. The reaction proceeds without racemization when optically active acids are used as the starting material. This method is operationally simple and provides the products in high purity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
N,N-Dimethylhydroxylamine hydrochloride, 99%