Merck
Fluorescence in situ hybridization (FISH) mapping of single copy genes on Trichomonas vaginalis chromosomes.
Molecular and biochemical parasitology (2011-01-05)
Zuzana Zubáčová, Vladimír Krylov, Jan Tachezy
RESUMEN
The highly repetitive nature of the Trichomonas vaginalis genome and massive expansion of various gene families has caused difficulties in genome assembly and has hampered genome mapping. Here, we adapted fluorescence in situ hybridization (FISH) for T. vaginalis, which is sensitive enough to detect single copy genes on metaphase chromosomes. Sensitivity of conventional FISH, which did not allow single copy gene detection in T. vaginalis, was increased by means of tyramide signal amplification. Two selected single copy genes, coding for serine palmitoyltransferase and tryptophanase, were mapped to chromosome I and II, respectively, and thus could be used as chromosome markers. This established protocol provides an amenable tool for the physical mapping of the T. vaginalis genome and other essential applications, such as development of genetic markers for T. vaginalis genotyping.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Apotryptophanase from Escherichia coli, soluble powder, 75-150 units/mg solid