Merck
Structural insight into nascent polypeptide chain-mediated translational stalling.
Science (New York, N.Y.) (2009-11-26)
Birgit Seidelt, C Axel Innis, Daniel N Wilson, Marco Gartmann, Jean-Paul Armache, Elizabeth Villa, Leonardo G Trabuco, Thomas Becker, Thorsten Mielke, Klaus Schulten, Thomas A Steitz, Roland Beckmann
RESUMEN
Expression of the Escherichia coli tryptophanase operon depends on ribosome stalling during translation of the upstream TnaC leader peptide, a process for which interactions between the TnaC nascent chain and the ribosomal exit tunnel are critical. We determined a 5.8 angstrom-resolution cryo-electron microscopy and single-particle reconstruction of a ribosome stalled during translation of the tnaC leader gene. The nascent chain was extended within the exit tunnel, making contacts with ribosomal components at distinct sites. Upon stalling, two conserved residues within the peptidyltransferase center adopted conformations that preclude binding of release factors. We propose a model whereby interactions within the tunnel are relayed to the peptidyltransferase center to inhibit translation. Moreover, we show that nascent chains adopt distinct conformations within the ribosomal exit tunnel.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Apotryptophanase from Escherichia coli, soluble powder, 75-150 units/mg solid