Merck
Hydrophobic derivatization of N-linked glycans for increased ion abundance in electrospray ionization mass spectrometry.
Journal of the American Society for Mass Spectrometry (2011-09-29)
S Hunter Walker, Laura M Lilley, Monica F Enamorado, Daniel L Comins, David C Muddiman
RESUMEN
A library of neutral, hydrophobic reagents was synthesized for use as derivatizing agents in order to increase the ion abundance of N-linked glycans in electrospray ionization mass spectrometry (ESI MS). The glycans are derivatized via hydrazone formation and are shown to increase the ion abundance of a glycan standard more than 4-fold. Additionally, the data show that the systematic addition of hydrophobic surface area to the reagent increases the glycan ion abundance, a property that can be further exploited in the analysis of glycans. The results of this study will direct the future synthesis of hydrophobic reagents for glycan analysis using the correlation between hydrophobicity and theoretical non-polar surface area calculation to facilitate the development of an optimum tag for glycan derivatization. The compatibility and advantages of this method are demonstrated by cleaving and derivatizing N-linked glycans from human plasma proteins. The ESI-MS signal for the tagged glycans are shown to be significantly more abundant, and the detection of negatively charged sialylated glycans is enhanced.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
A3 Glycan, from bovine, ≥90% (HPLC)