Merck
Detergent extraction of erythrocyte ghosts. Comparison of residues after cholate and Triton X-100 treatments.
Biochimica et biophysica acta (1976-06-04)
R Coleman, G Holdsworth, J B Finean
RESUMEN
1. Human erythrocyte ghosts were extracted with individual free and conjugated bile salts and, for comparison, with Triton X-100 under conditions approximating to physiological temperature, pH and tonicity. 2. Treatment with cholate, glycocholate, taurocholate, or with Triton X-100 gave lipid-depleted residues. These could still be seen as ghost-like profiles by phase contrast microscopy. Deopxycholate brought about complete membrane dissolutiom. 3. The cholate residue gave a trilamellar image by electron microscopy and in condensed form gave a smaller membrane repeat than untreated membranes. It had a polypeptide composition representing mainly integral proteins. 4. The Triton X-100 residue had a granular profile in the electron microscope and a polypeptide composition largely representing peripheral proteins.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%