Merck
Memory B Cell Activation Induced by Pertussis Booster Vaccination in Four Age Groups of Three Countries.
Frontiers in immunology (2022-06-10)
Pauline Versteegen, Alex-Mikael Barkoff, Marta Valente Pinto, Jan van de Kasteele, Aapo Knuutila, Sagida Bibi, Lia de Rond, Johanna Teräsjärvi, Katherine Sanders, Mary-Lène de Zeeuw-Brouwer, Raakel Luoto, Hinke Ten Hulscher, Elizabeth A Clutterbuck, Elisabeth A M Sanders, Jussi Mertsola, Guy A M Berbers, Qiushui He, Dominic F Kelly, Anne-Marie Buisman
RESUMEN
Immunogenicity of acellular pertussis (aP) vaccines is conventionally assessed by measuring antibody responses but antibody concentrations wane quickly after vaccination. Memory B cells, however, are critical in sustaining long-term protection and therefore may be an important factor when assessing pertussis immunity after vaccination. We studied pertussis specific memory B cell (re)activation induced by an aP booster vaccination in four different age groups within three countries. From a phase IV longitudinal interventional study, 268 participants across Finland, the Netherlands and the United Kingdom were included and received a 3-component pertussis booster vaccine: children (7-10y, n=53), adolescents (11-15y, n=66), young adults (20-34y, n=74), and older adults (60-70y, n=75). Memory B cells at baseline, day 28, and 1 year post-vaccination were measured by a pertussis toxin (Ptx), filamentous haemagglutinin (FHA), and pertactin (Prn) specific ELISpot assay. Antibody results measured previously were available for comparison. Furthermore, study participants were distributed into groups based on their baseline memory B cell frequencies, vaccine responses were monitored between these groups. Geometric mean (GM) memory B cell frequencies for pertussis antigens at baseline were low. At 28 days post-vaccination, these frequencies increased within each age group and were still elevated one year post-booster compared to baseline. Highest frequencies at day 28 were found within adolescents (GM: 5, 21, and 13, for Ptx, FHA and Prn, respectively) and lowest within older adults (GM: 2, 9, and 3, respectively). Moderate to strong correlations between memory B cell frequencies at day 28 and antibody concentrations at day 28 and 1 year were observed for Prn. Memory B cell frequencies > 1 per 100,000 PBMCs at baseline were associated with significantly higher memory responses after 28 days and 1 year. An aP booster vaccine (re)activated memory B cells in all age groups. Still elevated memory B cell frequencies after one year indicates enhanced immunological memory. However, antigen specific memory B cell activation seems weaker in older adults, which might reflect immunosenescence. Furthermore, the presence of circulating memory B cells at baseline positively affects memory B cell responses. This study was registered at www.clinicaltrialsregister.eu: No. 2016-003678-42.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Goat Anti-Human IgG, γ-Chain Specific Alkaline Phosphatase Conjugate, liquid, Calbiochem®