Merck
  • Dual Beneficial Effects of α-Spinasterol Isolated from Aster pseudoglehnii on Glucose Uptake in Skeletal Muscle Cells and Glucose-Stimulated Insulin Secretion in Pancreatic β-Cells.
Dual Beneficial Effects of α-Spinasterol Isolated from Aster pseudoglehnii on Glucose Uptake in Skeletal Muscle Cells and Glucose-Stimulated Insulin Secretion in Pancreatic β-Cells.
Plants (Basel, Switzerland) (2022-03-11)
Dahae Lee, Ji-Young Kim, Hak Cheol Kwon, Jaeyoung Kwon, Dae Sik Jang, Ki Sung Kang
RESUMEN
Herein, we determined whether α-Spinasterol, a stigmastane-type phytosterol isolated from Aster pseudoglehnii, potentially impacts glucose uptake and glucose-stimulated insulin secretion in skeletal muscle cells and pancreatic β-cells, respectively. We observed that A. pseudoglehnii and its fractions enhanced glucose uptake, with no toxic effects on C2C12 cells, with the n-hexane fraction exhibiting the most potent effect. α-Spinasterol, isolated from the n-hexane fraction, enhanced glucose uptake with no toxic effects on C2C12 cells. Additionally, α-Spinasterol increased the expression of associated proteins, including insulin receptor substrate-1, AMP-activated protein kinase, and glucose transporter type 4, as determined by Western blotting. Furthermore, α-Spinasterol enhanced insulin secretion in response to high glucose concentrations, with no toxic effects on INS-1 cells; this effect was superior to that demonstrated by gliclazide (positive control), commonly prescribed to treat type 2 diabetes (T2D). α-Spinasterol enhanced the expression of associated proteins, including insulin receptor substrate-2, peroxisome proliferator-activated receptor γ, and pancreatic and duodenal homeobox 1, as determined using Western blotting. The insulin secretory effect of α-Spinasterol was enhanced by a K+ channel blocker and L-type Ca2+ channel agonist and was suppressed by a K+ channel activator and L-type Ca2+ channel blocker. α-Spinasterol isolated from A. pseudoglehnii may improve hyperglycemia by improving glucose uptake into skeletal muscle cells and enhancing insulin secretion in pancreatic β-cells. Accordingly, α-Spinasterol could be a potential candidate for anti-T2D therapy.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-phospho-IRS-2 (Ser388) Antibody, from rabbit, purified by affinity chromatography
En este momento no podemos mostrarle ni los precios ni la disponibilidad