Merck
Microenvironmental IL1β promotes breast cancer metastatic colonisation in the bone via activation of Wnt signalling.
Nature communications (2019-11-05)
Rachel Eyre, Denis G Alférez, Angélica Santiago-Gómez, Kath Spence, James C McConnell, Claire Hart, Bruno M Simões, Diane Lefley, Claudia Tulotta, Joanna Storer, Austin Gurney, Noel Clarke, Mick Brown, Sacha J Howell, Andrew H Sims, Gillian Farnie, Penelope D Ottewell, Robert B Clarke
RESUMEN
Dissemination of tumour cells to the bone marrow is an early event in breast cancer, however cells may lie dormant for many years before bone metastases develop. Treatment for bone metastases is not curative, therefore new adjuvant therapies which prevent the colonisation of disseminated cells into metastatic lesions are required. There is evidence that cancer stem cells (CSCs) within breast tumours are capable of metastasis, but the mechanism by which these colonise bone is unknown. Here, we establish that bone marrow-derived IL1β stimulates breast cancer cell colonisation in the bone by inducing intracellular NFkB and CREB signalling in breast cancer cells, leading to autocrine Wnt signalling and CSC colony formation. Importantly, we show that inhibition of this pathway prevents both CSC colony formation in the bone environment, and bone metastasis. These findings establish that targeting IL1β-NFKB/CREB-Wnt signalling should be considered for adjuvant therapy to prevent breast cancer bone metastasis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Proteína DKK-1, recombinante humana, The Dickkopf related protein 1 (or Dkk1 protein) is the founding member of the Dickkopf family of proteins that includes Dkk1, 2, 3, 4 & a related protein, Soggy.