Merck
  • Development of potent dipeptide-type SARS-CoV 3CL protease inhibitors with novel P3 scaffolds: design, synthesis, biological evaluation, and docking studies.
Development of potent dipeptide-type SARS-CoV 3CL protease inhibitors with novel P3 scaffolds: design, synthesis, biological evaluation, and docking studies.
European journal of medicinal chemistry (2013-09-03)
Pillaiyar Thanigaimalai, Sho Konno, Takehito Yamamoto, Yuji Koiwai, Akihiro Taguchi, Kentaro Takayama, Fumika Yakushiji, Kenichi Akaji, Shen-En Chen, Aurash Naser-Tavakolian, Arne Schön, Ernesto Freire, Yoshio Hayashi
RESUMEN
We report the design and synthesis of a series of dipeptide-type inhibitors with novel P3 scaffolds that display potent inhibitory activity against SARS-CoV 3CLpro. A docking study involving binding between the dipeptidic lead compound 4 and 3CLpro suggested the modification of a structurally flexible P3 N-(3-methoxyphenyl)glycine with various rigid P3 moieties in 4. The modifications led to the identification of several potent derivatives, including 5c-k and 5n with the inhibitory activities (Ki or IC50) in the submicromolar to nanomolar range. Compound 5h, in particular, displayed the most potent inhibitory activity, with a Ki value of 0.006 μM. This potency was 65-fold higher than the potency of the lead compound 4 (Ki=0.39 μM). In addition, the Ki value of 5h was in very good agreement with the binding affinity (16 nM) observed in isothermal titration calorimetry (ITC). A SAR study around the P3 group in the lead 4 led to the identification of a rigid indole-2-carbonyl unit as one of the best P3 moieties (5c). Further optimization showed that a methoxy substitution at the 4-position on the indole unit was highly favorable for enhancing the inhibitory potency.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Mpro inhibitor 5h, ≥98% (HPLC)