Merck
  • Determination of the enantiomers of alpha-hydroxy- and alpha-amino acids in capillary electrophoresis with contactless conductivity detection.
Determination of the enantiomers of alpha-hydroxy- and alpha-amino acids in capillary electrophoresis with contactless conductivity detection.
Electrophoresis (2010-05-25)
Worapan Pormsila, Xiao Yang Gong, Peter C Hauser
RESUMEN
The enantiomers of the anions of five alpha-hydroxy acids, namely lactic acid, alpha-hydroxybutyric acid, 2-hydroxycaproic acid, 2-hydroxyoctanoic acid and 2-hydroxydecanoic acid, as well as the two alpha-amino acids aspartic acid and glutamic acid, were baseline separated and detected by CE with contactless conductivity detection. Vancomycin was employed as chiral selector and could be used with conductivity detection without having to resort to a partial filling protocol as needed when this reagent is used with UV absorbance measurements. The procedure was successfully applied to the determination of the lactic acid enantiomers in samples of milk and yogurt. Linearity was achieved in the concentration range of 10-500 micromol/L with good correlation coefficients (0.9993 and 0.9990 for L- and D-lactic acid, respectively). The LODs (3 S/N) for L- and D-lactic acid were determined as 2.8 and 2.4 micromol/L, respectively.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
(±)-2-Hydroxyoctanoic acid