Merck
  • Generation and characterization of a control and patient-derived human iPSC line containing the Hermansky Pudlak type 2 (HPS2) associated heterozygous compound mutation in AP3B1.
Generation and characterization of a control and patient-derived human iPSC line containing the Hermansky Pudlak type 2 (HPS2) associated heterozygous compound mutation in AP3B1.
Stem cell research (2021-06-29)
Cathelijn E M Aarts, Ellie Karampini, Tatjana Wüst, Steven Webbers, Eszter Varga, Judy Geissler, Jan Voorberg, Marieke von Lindern, Ruben Bierings, Emile van den Akker, Taco W Kuijpers
RESUMEN
Induced pluripotent stem cells (iPSCs) were generated from blood outgrowth endothelial cells (BOECs) obtained from a healthy donor and from a patient diagnosed with Hermansky Pudlak Syndrome type 2 (HPS2), caused by compound heterozygous AP3B1 mutations (c.177delA and c.1839-1842delTAGA). BOECs were reprogrammed with a hOKSM self-silencing polycistronic lentiviral vector, where the generated iPSCs showed normal karyotype, expression of pluripotency associated markers and in vitro spontaneous differentiation towards the three germ layers. The generated iPSCs can be used to study HPS2 pathophysiology and the basic functions of AP3B1 protein in different cell types.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, FITC conjugate, clone TRA-1-60, from mouse, FITC conjugate