Merck
Production of a fermented organic rice syrup with higher isomalto-oligosaccharide using Lactobacillus plantarum.
Food science and biotechnology (2018-09-29)
Hyo-Hee Kim, Won-Pyo Lee, Chang-Hwan Oh, Sung-Sik Yoon
RESUMEN
Isomalto-oligosaccharide (IMO) syrup was prepared from organic rice, in which Lactobacillus plantarum as a starter was inoculated to raise its purity and also produce a fermented rice beverage. Of the five strains of lactic acid bacteria tested, L. plantarum was preferentially selected in terms of a viable cells (7.3 × 108 colony forming unit (CFU/mL) and higher dry cell weight (13 mg/mL). The fermented syrup-based medium did not affect the growth of L. plantarum. As expected, the residual sugar content gradually decreased by 1.34% compared with the initial concentration. It was apparent that the residual sugars but not oligosaccharides were removed during the L. plantarum fermentation period. The production of lactic acid was the highest (8125.78 mg/kg) among the organic acids produced in the fermented IMO syrup.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido cítrico, ACS reagent, ≥99.5%