Merck
  • Generation of antibodies to Di- and trichloroacetylated proteins and immunochemical detection of protein adducts in rats treated with perchloroethene.
Generation of antibodies to Di- and trichloroacetylated proteins and immunochemical detection of protein adducts in rats treated with perchloroethene.
Chemical research in toxicology (1998-10-07)
A Pähler, G Birner, J Parker, W Dekant
RESUMEN
Antibodies directed against chemical specific protein modifications are valuable tools to detect and comparatively quantify protein modifications. Both Nepsilon-(dichloroacetyl)-L-lysine and Nepsilon-(trichloroacety)l-L-lysine have been detected as modified amino acids in liver and kidneys of rats treated with perchloroethene (PER) after proteolysis. These protein modifications are formed by the interaction of reactive metabolites formed from PER with proteins. In this study we developed monospecific antibodies to dichloroacetylated and to trichloroacetylated amino acids to detect modified proteins in the target organs of PER toxicity. These antibodies were prepared by immunization of rabbits with modified keyhole limpet hemocyanin (KLH) coupled with either the dichloroacetyl or trichloroacetyl moiety. Enzyme-linked immunosorbent assays (ELISA) indicated that the polyclonal rabbit sera recognized dichloroacetylated or trichloroacetylated rabbit serum albumin (RSA), but not unmodified protein. Therefore, we further purified rabbit antisera on either Nepsilon-(dichloroacetyl)-L-lysine or Nepsilon-(trichloroacetyl)-L-lysine immobilized to immunoaffinity columns to obtain monospecific antibodies. The potential of these antibodies in the detection of di- and trichloroacetylated proteins and their selectivity for the desired dichloroacetyl or trichloroacetyl group was demonstrated in competitive enzme-linked immunosorbent assays with several structurally related compounds. Anti-dichloroacetyl (anti-DCA) antibody binding to dichloroacetylated RSA was inhibited by Nepsilon-(dichloroacetyl)-L-lysine with an IC50 value of 150 microM whereas inhibition by Nepsilon-(monochloroacetyl)-L-lysine and Nepsilon-(trichloroacetyl)-L-lysine showed an IC50 value of 100 mM. The binding of the anti-trichloroacetyl (anti-TCA) antibody to trichloroacetylated RSA was inhibited by Nepsilon-(dichloroacetyl)-L-lysine with an IC50 value of 80 mM. The inhibition by Nepsilon-(trichloroacetyl)-L-lysine was again 3 orders of magnitude stronger resulting in an IC50 value of 90 microM. Nepsilon-(acetyl)-L-lysine and unmodified RSA did not effect antibody binding to the chemically modified antigen. The antibodies were also successfully applied to detect modified proteins in subcellular fractions of liver and kidney from PER treated rats demonstrated in immunoblot. Protein adduct formation from different PER metabolism pathways was confirmed by the observation that the majority of dichloroacetylated proteins were located in kidney mitochondria and trichloroacetylated proteins were located in liver microsomes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nε-Acetyl-L-lysine