Merck
The modular phosphorylation and activation of p70s6k.
FEBS letters (1997-06-23)
N Pullen, G Thomas
RESUMEN
The activation of p70s6k is accompanied by a complex series of phosphorylation events. In this review we propose a model of activation which divides p70s6k into four functional modules that cooperate in leading to full enzyme activity. In the light of the model, we suggest how candidate effectors of p70s6k activation might function by directing the phosphorylation of specific sites.