Merck
Loss of neuronal Miro1 disrupts mitophagy and induces hyperactivation of the integrated stress response.
The EMBO journal (2021-06-22)
Guillermo López-Doménech, Jack H Howden, Christian Covill-Cooke, Corinne Morfill, Jigna V Patel, Roland Bürli, Damian Crowther, Nicol Birsa, Nicholas J Brandon, Josef T Kittler
RESUMEN
Clearance of mitochondria following damage is critical for neuronal homeostasis. Here, we investigate the role of Miro proteins in mitochondrial turnover by the PINK1/Parkin mitochondrial quality control system in vitro and in vivo. We find that upon mitochondrial damage, Miro is promiscuously ubiquitinated on multiple lysine residues. Genetic deletion of Miro or block of Miro1 ubiquitination and subsequent degradation lead to delayed translocation of the E3 ubiquitin ligase Parkin onto damaged mitochondria and reduced mitochondrial clearance in both fibroblasts and cultured neurons. Disrupted mitophagy in vivo, upon post-natal knockout of Miro1 in hippocampus and cortex, leads to a dramatic increase in mitofusin levels, the appearance of enlarged and hyperfused mitochondria and hyperactivation of the integrated stress response (ISR). Altogether, our results provide new insights into the central role of Miro1 in the regulation of mitochondrial homeostasis and further implicate Miro1 dysfunction in the pathogenesis of human neurodegenerative disease.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti--actina antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-fosfo-ubiquitina (Ser65), from rabbit, purified by affinity chromatography
En este momento no podemos mostrarle ni los precios ni la disponibilidad